Anti-FOXD3 Rat Monoclonal Antibody [clone: 800602]

Supplier: R & D Systems
MAB5090
102885-890EA 577.02 USD
102885-890
Anti-FOXD3 Rat Monoclonal Antibody [clone: 800602]
Antibodies
Type: Primary
Antigen Symbol: FOXD3
Antigen Name: forkhead box D3
Antigen Synonyms: CWH3, Genesis, Hfh2
Clonality: Monoclonal
Clone: 800602
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR