Anti-APOH Mouse Monoclonal Antibody [clone: 517038]

Supplier: R & D Systems
MAB5087
220021-087EA 594.37 USD
220021-087
Anti-APOH Mouse Monoclonal Antibody [clone: 517038]
Antibodies
Type: Primary
Antigen: APOH
Clonality: Monoclonal
Clone: 517038
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR