Anti-CCL23 Mouse Monoclonal Antibody [clone: 103952]

Supplier: R & D Systems
MAB508
220021-086EA 772.25 USD
220021-086
Anti-CCL23 Mouse Monoclonal Antibody [clone: 103952]
Antibodies
Type: Primary
Antigen: CCL23
Clonality: Monoclonal
Clone: 103952
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR