Anti-RELA Rat Monoclonal Antibody [clone: 465003]

Supplier: R & D Systems
MAB50781
102885-888EA 577.02 USD
102885-888
Anti-RELA Rat Monoclonal Antibody [clone: 465003]
Antibodies
Type: Primary
Antigen: RELA
Clonality: Monoclonal
Clone: 465003
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR