Anti-RELA Mouse Monoclonal Antibody [clone: 532301]

Supplier: R & D Systems
MAB5078
220021-085EA 647.88 USD
220021-085
Anti-RELA Mouse Monoclonal Antibody [clone: 532301]
Antibodies
Type: Primary
Antigen: RELA
Clonality: Monoclonal
Clone: 532301
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR