Anti-ACP1 Mouse Monoclonal Antibody [clone: 475417]

Supplier: R & D Systems
MAB5075
220021-084EA 594.37 USD
220021-084
Anti-ACP1 Mouse Monoclonal Antibody [clone: 475417]
Antibodies
Type: Primary
Antigen: ACP1
Clonality: Monoclonal
Clone: 475417
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR