Anti-PPP4C Mouse Monoclonal Antibody [clone: 501517]

Supplier: R & D Systems
MAB5074
220021-083EA 577.02 USD
220021-083
Anti-PPP4C Mouse Monoclonal Antibody [clone: 501517]
Antibodies
Type: Primary
Antigen: PPP4C
Clonality: Monoclonal
Clone: 501517
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR