Anti-HS6ST1 Mouse Monoclonal Antibody [clone: 584308]

Supplier: R & D Systems
MAB5057
220021-079EA 577.02 USD
220021-079
Anti-HS6ST1 Mouse Monoclonal Antibody [clone: 584308]
Antibodies
Type: Primary
Antigen: HS6ST1
Clonality: Monoclonal
Clone: 584308
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR