Anti-BMPR1B Mouse Monoclonal Antibody [clone: 477914]

Supplier: R & D Systems
MAB5051
220021-078EA 642.09 USD
220021-078
Anti-BMPR1B Mouse Monoclonal Antibody [clone: 477914]
Antibodies
Type: Primary
Antigen: BMPR1B
Clonality: Monoclonal
Clone: 477914
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR