Anti-CD63 Mouse Monoclonal Antibody [clone: 460305]

Supplier: R & D Systems
MAB5048
220021-076EA 616.06 USD
220021-076
Anti-CD63 Mouse Monoclonal Antibody [clone: 460305]
Antibodies
Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: 460305
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR