Anti-PRSS22 Mouse Monoclonal Antibody [clone: 725445]

Supplier: R & D Systems
MAB5047
102885-886EA 577.02 USD
102885-886
Anti-PRSS22 Mouse Monoclonal Antibody [clone: 725445]
Antibodies
Type: Primary
Antigen: PRSS22
Clonality: Monoclonal
Clone: 725445
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR