Anti-MSX1 Mouse Monoclonal Antibody [clone: 801601]

Supplier: R & D Systems
MAB5045
102885-884EA 577.02 USD
102885-884
Anti-MSX1 Mouse Monoclonal Antibody [clone: 801601]
Antibodies
Type: Primary
Antigen: MSX1
Clonality: Monoclonal
Clone: 801601
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR