Anti-FOXC2 Mouse Monoclonal Antibody [clone: 665819]

Supplier: R & D Systems
MAB5044
102885-882EA 594.37 USD
102885-882
Anti-FOXC2 Mouse Monoclonal Antibody [clone: 665819]
Antibodies
Type: Primary
Antigen Symbol: FOXC2
Antigen Name: forkhead box C2
Antigen Synonyms: FKHL14, LD, MFH-1, MFH1
Clonality: Monoclonal
Clone: 665819
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR