Anti-TBX2 Rat Monoclonal Antibody [clone: 393720]

Supplier: R & D Systems
MAB50401
220021-074EA 577.02 USD
220021-074
Anti-TBX2 Rat Monoclonal Antibody [clone: 393720]
Antibodies
Type: Primary
Antigen: TBX2
Clonality: Monoclonal
Clone: 393720
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR