Anti-TBX2 Rat Monoclonal Antibody [clone: 393725]

Supplier: R & D Systems
MAB5040
220021-073EA 561.11 USD
220021-073
Anti-TBX2 Rat Monoclonal Antibody [clone: 393725]
Antibodies
Type: Primary
Antigen: TBX2
Clonality: Monoclonal
Clone: 393725
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR