Anti-TNFA Mouse Monoclonal Antibody [clone: 569826]

Supplier: R & D Systems
MAB5035
102885-880EA 763.57 USD
102885-880
Anti-TNFA Mouse Monoclonal Antibody [clone: 569826]
Antibodies
Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: 569826
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR