Anti-IL33 (Propeptide) Rat Monoclonal Antibody [clone: 518017]

Supplier: R & D Systems
MAB5010
220021-068EA 577.02 USD
220021-068
Anti-IL33 (Propeptide) Rat Monoclonal Antibody [clone: 518017]
Antibodies
Type: Primary
Antigen: IL33 (Propeptide)
Clonality: Monoclonal
Clone: 518017
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR