Anti-IL1B Mouse Monoclonal Antibody [clone: 38139]

Supplier: R & D Systems
MAB501
220021-067EA 772.25 USD
220021-067
Anti-IL1B Mouse Monoclonal Antibody [clone: 38139]
Antibodies
Type: Primary
Antigen: IL1B
Clonality: Monoclonal
Clone: 38139
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR