Anti-CITED2 Rat Monoclonal Antibody [clone: 477226]

Supplier: R & D Systems
MAB5005
102885-874EA 594.37 USD
102885-874
Anti-CITED2 Rat Monoclonal Antibody [clone: 477226]
Antibodies
Type: Primary
Antigen: CITED2
Clonality: Monoclonal
Clone: 477226
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR