Anti-IL1A Mouse Monoclonal Antibody [clone: 59015]

Supplier: R & D Systems
MAB500
220021-065EA 772.25 USD
220021-065
Anti-IL1A Mouse Monoclonal Antibody [clone: 59015]
Antibodies
Type: Primary
Antigen: IL1A
Clonality: Monoclonal
Clone: 59015
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR