Anti-RAD50 Mouse Monoclonal Antibody [clone: 684945]

Supplier: R & D Systems
MAB4996
102885-870EA 561.11 USD
102885-870
Anti-RAD50 Mouse Monoclonal Antibody [clone: 684945]
Antibodies
Type: Primary
Antigen: RAD50
Clonality: Monoclonal
Clone: 684945
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR