Anti-LRRC4 Mouse Monoclonal Antibody [clone: 701424]

Supplier: R & D Systems
MAB4995
102885-866EA 594.37 USD
102885-866
Anti-LRRC4 Mouse Monoclonal Antibody [clone: 701424]
Antibodies
Type: Primary
Antigen: LRRC4
Clonality: Monoclonal
Clone: 701424
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR