Anti-HSPB8 Mouse Monoclonal Antibody [clone: 520817]

Supplier: R & D Systems
MAB4987
220021-062EA 594.37 USD
220021-062
Anti-HSPB8 Mouse Monoclonal Antibody [clone: 520817]
Antibodies
Type: Primary
Antigen: HSPB8
Clonality: Monoclonal
Clone: 520817
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR