Anti-STK17A Mouse Monoclonal Antibody [clone: 684324]

Supplier: R & D Systems
MAB4985
102885-864EA 577.02 USD
102885-864
Anti-STK17A Mouse Monoclonal Antibody [clone: 684324]
Antibodies
Type: Primary
Antigen: STK17A
Clonality: Monoclonal
Clone: 684324
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR