Anti-SOCS2 Mouse Monoclonal Antibody [clone: 517206]

Supplier: R & D Systems
MAB4979
220021-061EA 594.37 USD
220021-061
Anti-SOCS2 Mouse Monoclonal Antibody [clone: 517206]
Antibodies
Type: Primary
Antigen: SOCS2
Clonality: Monoclonal
Clone: 517206
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR