Anti-PROS1 Rat Monoclonal Antibody [clone: 818002]

Supplier: R & D Systems
MAB4976
102885-862EA 633.42 USD
102885-862
Anti-PROS1 Rat Monoclonal Antibody [clone: 818002]
Antibodies
Type: Primary
Antigen: PROS1
Clonality: Monoclonal
Clone: 818002
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR