Anti-TPBG Mouse Monoclonal Antibody [clone: 524744]

Supplier: R & D Systems
MAB49751
220021-060EA 623.29 USD
220021-060
Anti-TPBG Mouse Monoclonal Antibody [clone: 524744]
Antibodies
Type: Primary
Antigen: TPBG
Clonality: Monoclonal
Clone: 524744
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR