Anti-TPBG Mouse Monoclonal Antibody [clone: 524731]

Supplier: R & D Systems
MAB4975
220021-059EA 655.11 USD
220021-059
Anti-TPBG Mouse Monoclonal Antibody [clone: 524731]
Antibodies
Type: Primary
Antigen: TPBG
Clonality: Monoclonal
Clone: 524731
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR