Anti-LEPR Rat Monoclonal Antibody [clone: 55305]

Supplier: R & D Systems
MAB497
220021-057EA 763.57 USD
220021-057
Anti-LEPR Rat Monoclonal Antibody [clone: 55305]
Antibodies
Type: Primary
Antigen: LEPR
Clonality: Monoclonal
Clone: 55305
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR