Anti-ADGRB1 Mouse Monoclonal Antibody [clone: 542515]

Supplier: R & D Systems
MAB4969
220021-056EA 594.37 USD
220021-056
Anti-ADGRB1 Mouse Monoclonal Antibody [clone: 542515]
Antibodies
Type: Primary
Antigen: ADGRB1
Clonality: Monoclonal
Clone: 542515
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR