Anti-OSM Rat Monoclonal Antibody [clone: 157210]

Supplier: R & D Systems
MAB495
220021-055EA 763.57 USD
220021-055
Anti-OSM Rat Monoclonal Antibody [clone: 157210]
Antibodies
Type: Primary
Antigen: OSM
Clonality: Monoclonal
Clone: 157210
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR