Anti-APLP2 Mouse Monoclonal Antibody [clone: 39772]

Supplier: R & D Systems
MAB49451
102885-858EA 655.11 USD
102885-858
Anti-APLP2 Mouse Monoclonal Antibody [clone: 39772]
Antibodies
Type: Primary
Antigen: APLP2
Clonality: Monoclonal
Clone: 39772
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR