Anti-REG1A Rat Monoclonal Antibody [clone: 431211]

Supplier: R & D Systems
MAB49371
102885-856EA 566.89 USD
102885-856
Anti-REG1A Rat Monoclonal Antibody [clone: 431211]
Antibodies
Type: Primary
Antigen: REG1A
Clonality: Monoclonal
Clone: 431211
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR