Anti-REG1A Rat Monoclonal Antibody [clone: 431202]

Supplier: R & D Systems
MAB4937
220021-054EA 594.37 USD
220021-054
Anti-REG1A Rat Monoclonal Antibody [clone: 431202]
Antibodies
Type: Primary
Antigen: REG1A
Clonality: Monoclonal
Clone: 431202
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR