Anti-NEDD8 Mouse Monoclonal Antibody [clone: 584606]

Supplier: R & D Systems
MAB4936
102885-854EA 561.11 USD
102885-854
Anti-NEDD8 Mouse Monoclonal Antibody [clone: 584606]
Antibodies
Type: Primary
Antigen Symbol: NEDD8
Antigen Name: neural precursor cell expressed, developmentally down-regulated 8
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 584606
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR