Anti-FOSL1 Mouse Monoclonal Antibody [clone: 496106]

Supplier: R & D Systems
MAB4935
102885-852EA 577.02 USD
102885-852
Anti-FOSL1 Mouse Monoclonal Antibody [clone: 496106]
Antibodies
Type: Primary
Antigen Symbol: FOSL1
Antigen Name: FOS-like antigen 1
Antigen Synonyms: FRA, FRA1, fra-1
Clonality: Monoclonal
Clone: 496106
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR