Anti-STAT3 Mouse Monoclonal Antibody [clone: 788335]

Supplier: R & D Systems
MAB4934
102885-850EA 594.37 USD
102885-850
Anti-STAT3 Mouse Monoclonal Antibody [clone: 788335]
Antibodies
Type: Primary
Antigen: STAT3
Clonality: Monoclonal
Clone: 788335
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR