Anti-VEGFA Rat Monoclonal Antibody [clone: 39917]

Supplier: R & D Systems
MAB493
220021-052EA 763.57 USD
220021-052
Anti-VEGFA Rat Monoclonal Antibody [clone: 39917]
Antibodies
Type: Primary
Antigen: VEGFA
Clonality: Monoclonal
Clone: 39917
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR