Anti-ERMAP Mouse Monoclonal Antibody [clone: 767505]

Supplier: R & D Systems
MAB49281
102885-848EA 577.02 USD
102885-848
Anti-ERMAP Mouse Monoclonal Antibody [clone: 767505]
Antibodies
Type: Primary
Antigen: ERMAP
Clonality: Monoclonal
Clone: 767505
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR