Anti-ERMAP Mouse Monoclonal Antibody [clone: 767504]

Supplier: R & D Systems
MAB4928
102885-846EA 561.11 USD
102885-846
Anti-ERMAP Mouse Monoclonal Antibody [clone: 767504]
Antibodies
Type: Primary
Antigen: ERMAP
Clonality: Monoclonal
Clone: 767504
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR