Anti-PON1 Rat Monoclonal Antibody [clone: 453223]

Supplier: R & D Systems
MAB4926
220021-051EA 577.02 USD
220021-051
Anti-PON1 Rat Monoclonal Antibody [clone: 453223]
Antibodies
Type: Primary
Antigen: PON1
Clonality: Monoclonal
Clone: 453223
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR