Anti-ENPP7 Mouse Monoclonal Antibody [clone: 560514]

Supplier: R & D Systems
MAB4924
102885-844EA 594.37 USD
102885-844
Anti-ENPP7 Mouse Monoclonal Antibody [clone: 560514]
Antibodies
Type: Primary
Antigen: ENPP7
Clonality: Monoclonal
Clone: 560514
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR