Anti-SEZ6L2 Rat Monoclonal Antibody [clone: 529805]

Supplier: R & D Systems
MAB4916
220021-050EA 577.02 USD
220021-050
Anti-SEZ6L2 Rat Monoclonal Antibody [clone: 529805]
Antibodies
Type: Primary
Antigen Symbol: SEZ6L2
Antigen Name: seizure related 6 homolog like 2
Antigen Synonyms: AI835913, AW121566, BSRP-A, Psk1
Clonality: Monoclonal
Clone: 529805
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR