Anti-CR2 Mouse Monoclonal Antibody [clone: 544408]

Supplier: R & D Systems
MAB4909
102885-840EA 616.06 USD
102885-840
Anti-CR2 Mouse Monoclonal Antibody [clone: 544408]
Antibodies
Type: Primary
Antigen: CR2
Clonality: Monoclonal
Clone: 544408
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR