Anti-ADAM22 Mouse Monoclonal Antibody [clone: 49885]

Supplier: R & D Systems
MAB49081
220021-049EA 577.02 USD
220021-049
Anti-ADAM22 Mouse Monoclonal Antibody [clone: 49885]
Antibodies
Type: Primary
Antigen: ADAM22
Clonality: Monoclonal
Clone: 49885
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR