Anti-RYK Mouse Monoclonal Antibody [clone: 484804]

Supplier: R & D Systems
MAB4907
102885-838EA 594.37 USD
102885-838
Anti-RYK Mouse Monoclonal Antibody [clone: 484804]
Antibodies
Type: Primary
Antigen: RYK
Clonality: Monoclonal
Clone: 484804
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR