Anti-SCARA5 Mouse Monoclonal Antibody [clone: 673527]

Supplier: R & D Systems
MAB4900
102885-836EA 594.37 USD
102885-836
Anti-SCARA5 Mouse Monoclonal Antibody [clone: 673527]
Antibodies
Type: Primary
Antigen: SCARA5
Clonality: Monoclonal
Clone: 673527
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR