Anti-LRRTM3 Mouse Monoclonal Antibody [clone: 568318]

Supplier: R & D Systems
MAB4898
102885-832EA 561.11 USD
102885-832
Anti-LRRTM3 Mouse Monoclonal Antibody [clone: 568318]
Antibodies
Type: Primary
Antigen: LRRTM3
Clonality: Monoclonal
Clone: 568318
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR