Anti-ADGRE2 Mouse Monoclonal Antibody [clone: 49425]

Supplier: R & D Systems
MAB4894
220021-048EA 642.09 USD
220021-048
Anti-ADGRE2 Mouse Monoclonal Antibody [clone: 49425]
Antibodies
Type: Primary
Antigen: ADGRE2
Clonality: Monoclonal
Clone: 49425
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR