Anti-CLEC10A Mouse Monoclonal Antibody [clone: 744812]

Supplier: R & D Systems
MAB48881
102885-830EA 600.15 USD
102885-830
Anti-CLEC10A Mouse Monoclonal Antibody [clone: 744812]
Antibodies
Type: Primary
Antigen: CLEC10A
Clonality: Monoclonal
Clone: 744812
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR